Bicyclo[2.2.1]heptane-2-carbaldehyde 95%

Supplier: Ambeed
Danger

A705369-100MG A705369-5G
76727-728EA 51 USD
76727-728 77880-464
Bicyclo[2.2.1]heptane-2-carbaldehyde 95%
Bicyclo[2.2.1]heptane-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₂O
MDL Number: MFCD01318318
CAS Number: 19396-83-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR