4-Methoxybenzaldehyde dimethyl acetal 97%

Supplier: Ambeed
Warning

Synonyms: p-(Dimethoxymethyl)anisole, P-anisaldehyde dimethyl acetal, p-Anisaldehyde dimethyl acetal, 1-(Dimethoxymethyl)-4-methoxybenzene, 4-Methoxy-α,α-dimethoxytoluene

A705123-100G A705123-25G A705123-500G A705123-5G
76727-714EA 52 USD
76727-714 76727-716 76727-718 76727-720
4-Methoxybenzaldehyde dimethyl acetal 97%
4-Methoxybenzaldehyde dimethyl acetal

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD00036507
CAS Number: 2186-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR