Methyl 4-hydroxy-2-methyl-2H-thieno[2,3-e][1,2]thiazine-3-carboxylate 1,1-dioxide 95%

Supplier: Ambeed
Warning

A699884-1G A699884-25G A699884-5G
76727-682EA 29.71 USD
76727-682 76727-684 76727-686
Methyl 4-hydroxy-2-methyl-2H-thieno[2,3-e][1,2]thiazine-3-carboxylate 1,1-dioxide 95%
Methyl 4-hydroxy-2-methyl-2H-thieno[2,3-e][1,2]thiazine-3-carboxylate 1,1-dioxide

Precaución: for Research Use Only

Formula: C₉H₉NO₅S₂
MW: 275.30 g/mol
MDL Number: MFCD08460400
CAS Number: 59804-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR