N-(2-Hydroxyethyl)succinimide 97%

Supplier: Ambeed
Warning

A699290-100G A699290-25G A699290-500G A699290-5G
76727-674EA 109.28 USD
76727-674 76727-676 76727-678 76727-680
N-(2-Hydroxyethyl)succinimide 97%
N-(2-Hydroxyethyl)succinimide

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.14 g/mol
MDL Number: MFCD00078332
CAS Number: 18190-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR