Trimethoxy(p-tolyl)silane 95%

Supplier: Ambeed
Warning

A698976-1G A698976-25G A698976-5G
76727-668EA 21 USD
76727-668 76727-670 76727-672
Trimethoxy(p-tolyl)silane 95%
Trimethoxy(p-tolyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₃Si
MDL Number: MFCD00048020
CAS Number: 17873-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR