2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(1,3,2-dioxaborinan-2-yl)benzoate

A697409-1G A697409-250MG A697409-5G
76727-660EA 43.5 USD
76727-660 76727-662 76727-664
2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester 96%
2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BO₄
MW: 234.0561 g/mol
MDL Number: MFCD06659932
CAS Number: 850567-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR