3-Chloro-5-ethynylpyridine 95%

Supplier: Ambeed
Warning

A693989-1G A693989-250MG
76727-648EA 145.35 USD
76727-648 76727-650
3-Chloro-5-ethynylpyridine 95%
3-Chloro-5-ethynylpyridine

Precaución: for Research Use Only

Formula: C₇H₄ClN
MW: 137.57 g/mol
MDL Number: MFCD09832884
CAS Number: 329202-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR