N2,N4-Diethyl-6-methoxy-1,3,5-triazine-2,4-diamine 97%

Supplier: Ambeed
Warning

A692646-100MG A692646-1G A692646-250MG A692646-5G
76727-640EA 13.8 USD
76727-640 76727-642 76727-644 76727-646
N2,N4-Diethyl-6-methoxy-1,3,5-triazine-2,4-diamine 97%
N2,N4-Diethyl-6-methoxy-1,3,5-triazine-2,4-diamine

Precaución: for Research Use Only

Formula: C₈H₁₅N₅O
MW: 197.23 g/mol
MDL Number: MFCD01311011
CAS Number: 673-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR