6-Methylthiochromane-7-sulfonamide 1,1-dioxide 98%

Supplier: Ambeed
Warning

A692089-25MG A692089-1G A692089-250MG A692089-100MG
76727-638EA 42 USD
76727-638 77880-439 77880-440 77880-438
6-Methylthiochromane-7-sulfonamide 1,1-dioxide 98%
6-Methylthiochromane-7-sulfonamide 1,1-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S₂
MDL Number: MFCD00079454
CAS Number: 1084-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR