Potassium sulfamate 98%

Supplier: Ambeed
Warning

A691148-100G A691148-500G
76727-634EA 40 USD
76727-634 76727-636
Potassium sulfamate 98%
Potassium sulfamate

Precaución: for Research Use Only

Formula: H₃NO₃S
MDL Number: MFCD00059941
CAS Number: 13823-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR