1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate
A690975-100G
A690975-25G
76727-630EA
53.05
USD
76727-630
76727-632
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate 98%
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate
Precaución: for Research Use Only
Formula:
C₆H₄Na₂O₈S₂·1H₂O MW: 332.22 g/mol |
MDL Number:
MFCD00149531 CAS Number: 270573-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...