N-Ethyl-p-toluenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-4-methylbenzenesulphonamide, p-Tolueneethylsulphonamide, N-Ethyltoluene-4-sulphonamide, N-Tosylethylamine

A690478-1000G A690478-100G A690478-25G A690478-500G
76727-620EA 82.76 USD
76727-620 76727-622 76727-624 76727-626
N-Ethyl-p-toluenesulfonamide 98%
N-Ethyl-p-toluenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
MDL Number: MFCD00048511
CAS Number: 80-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR