N-Methyl-N-methoxydifluoroacetamide 97%

Supplier: Ambeed
Danger

A689946-25G A689946-5G
76727-612EA 141 USD
76727-612 76727-614
N-Methyl-N-methoxydifluoroacetamide 97%
N-Methyl-N-methoxydifluoroacetamide

Precaución: for Research Use Only

Formula: C₄H₇F₂NO₂
MDL Number: MFCD06248152
CAS Number: 142492-01-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR