3-(4-Pyridyl)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyridin-4-yl)benzaldehyde, 3-Pyrid-4-ylbenzaldehyde

A686640-1G A686640-5G
76727-606EA 20 USD
76727-606 76727-608
3-(4-Pyridyl)benzaldehyde 95%
3-(4-Pyridyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MDL Number: MFCD02684105
CAS Number: 208190-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR