Norcamphor 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Oxonorbornane, Bicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]-2-heptanone, 2,5-Methanocyclohexanone, 2-Norbornanone
A684819-1G
A684819-25G
A684819-5G
76727-598EA
13.8
USD
76727-598
76727-600
76727-602
Norcamphor 98%
Norcamphor
Precaución: for Research Use Only
Formula:
C₇H₁₀O MW: 110.16 g/mol |
MDL Number:
MFCD00074823 CAS Number: 497-38-1 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...