Sodium 3-(ethyl(3-methoxyphenyl)amino)-2-hydroxypropane-1-sulfonate 98%

Supplier: Ambeed
Warning

A680953-100MG A680953-1G A680953-250MG A680953-5G
76727-580EA 12.74 USD
76727-580 76727-582 76727-584 76727-586
Sodium 3-(ethyl(3-methoxyphenyl)amino)-2-hydroxypropane-1-sulfonate 98%
Sodium 3-(ethyl(3-methoxyphenyl)amino)-2-hydroxypropane-1-sulfonate

Precaución: for Research Use Only

Formula: C₁₂H₁₈NNaO₅S
MW: 311.33 g/mol
MDL Number: MFCD00152071
CAS Number: 82692-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR