(R)-2-((tert-Butoxycarbonyl)amino)-2-(3-chlorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A680599-1G A680599-250MG
76727-576EA 95 USD
76727-576 76727-578
(R)-2-((tert-Butoxycarbonyl)amino)-2-(3-chlorophenyl)acetic acid 95%
(R)-2-((tert-Butoxycarbonyl)amino)-2-(3-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO₄
MDL Number: MFCD07371716
CAS Number: 926641-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR