Oxamide 97%

Supplier: Ambeed
Danger

A679587-1G A679587-100G
76727-566EA 5.31 USD
76727-566 77573-198
Oxamide 97%
Oxamide

Precaución: for Research Use Only

Formula: C₂H₄N₂O₂
MW: 88.07 g/mol
MDL Number: MFCD00008007
CAS Number: 471-46-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR