Potassium formate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Formic acid potassium salt
A679302-1000G
A679302-100G
A679302-500G
76727-560EA
28.65
USD
76727-560
76727-562
76727-564
Potassium formate 97%
Potassium formate
Precaución: for Research Use Only
Formula:
CHKO₂ MW: 84.12 g/mol |
MDL Number:
MFCD00013100 CAS Number: 590-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...