4-(5-Fluoro-1H-indol-3-yl)butanoic acid 97%

Supplier: Ambeed
Warning

A679176-1G A679176-250MG A679176-5G
76727-554EA 83 USD
76727-554 76727-556 76727-558
4-(5-Fluoro-1H-indol-3-yl)butanoic acid 97%
4-(5-Fluoro-1H-indol-3-yl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂FNO₂
MW: 221.22 g/mol
MDL Number: MFCD03839847
CAS Number: 319-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR