2,2':5',2''-Terthiophene-5-carboxaldehyde 98%

Supplier: Ambeed
Warning

A672963-1G A672963-250MG A672963-5G A672963-25G
76727-532EA 33 USD
76727-532 76727-534 76727-536 77873-076
2,2':5',2''-Terthiophene-5-carboxaldehyde 98%
2,2':5',2''-Terthiophene-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈OS₃
MDL Number: MFCD00115181
CAS Number: 7342-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR