Methyl-5-methoxysalicylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-5-methoxybenzoate

A672771-1G A672771-5G
76727-528EA 9 USD
76727-528 76727-530
Methyl-5-methoxysalicylate 97%
Methyl 5-methoxysalicylate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
MDL Number: MFCD00017185
CAS Number: 2905-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR