tert-Butyl cyclohexyl(3-hydroxypropyl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A671659-1G
A671659-250MG
76727-524EA
101
USD
76727-524
76727-526
tert-Butyl cyclohexyl(3-hydroxypropyl)carbamate 97%
tert-Butyl cyclohexyl(3-hydroxypropyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₄H₂₇NO₃ |
MDL Number:
MFCD09037944 CAS Number: 266690-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...