3-Phenoxybenzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3-Phenoxybenzenecarbaldehyde, m-Phenoxybenzenecarbaldehyde

A671060-100G A671060-10G A671060-25G A671060-500G A671060-5G
76727-514EA 39.26 USD
76727-514 76727-516 76727-518 76727-520 76727-522
3-Phenoxybenzaldehyde 95%
3-Phenoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD00003353
CAS Number: 39515-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR