N-Isopropylaniline 98%
Supplier: Ambeed
|
Warning
|
A669892-100G
A669892-10G
A669892-25G
A669892-500G
76727-508EA
41
USD
76727-508
76727-510
76727-512
77880-402
N-Isopropylaniline 98%
N-Isopropylaniline
Precaución: for Research Use Only
Formula:
C₆H₅NHCH(CH₃)₂ |
MDL Number:
MFCD00026347 CAS Number: 768-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...