Boc-D-Thi-OH 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(thiophen-2-yl)propanoic acid, Boc-D-β-(2-Thienyl)-Ala-OH

A669835-1G A669835-250MG A669835-5G
76727-502EA 29 USD
76727-502 76727-504 76727-506
Boc-D-Thi-OH 95%
Boc-D-Thi-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₄S
MW: 271.34 g/mol
MDL Number: MFCD00065591
CAS Number: 78452-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR