Phthalimidoacetone 95%

Supplier: Ambeed
Warning

A667784-100G A667784-10G A667784-1G A667784-25G A667784-5G
76727-488EA 207 USD
76727-488 76727-490 76727-492 76727-494 76727-496
Phthalimidoacetone 95%
Phthalimidoacetone

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MDL Number: MFCD00088346
CAS Number: 3416-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR