2-(Methoxycarbonyl)nicotinic acid 96%

Supplier: Ambeed
Warning

A666724-1G A666724-250MG A666724-5G
76727-476EA 50 USD
76727-476 76727-478 76727-480
2-(Methoxycarbonyl)nicotinic acid 96%
2-(Methoxycarbonyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00111560
CAS Number: 24195-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR