O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: PFBHA.HCl

A662535-100MG A662535-1G A662535-250MG
76727-446EA 10.61 USD
76727-446 76727-448 76727-450
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

Precaución: for Research Use Only

Formula: C₇H₄F₅NO
MW: 249.57 g/mol
MDL Number: MFCD00012953
CAS Number: 57981-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR