Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate 98%

Supplier: Ambeed
Warning

Synonyms: S-(+)-Methoprene, Prop-2-yl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate

A662508-1G A662508-250MG A662508-5G
76727-440EA 33.95 USD
76727-440 76727-442 76727-444
Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate 98%
Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate

Precaución: for Research Use Only

Formula: C₁₉H₃₄O₃
MW: 310.471 g/mol
MDL Number: MFCD00867612
CAS Number: 65733-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR