Octaphenylcyclotetrasiloxane 98%

Supplier: Ambeed
Warning

Synonyms: 2,2,4,4,6,6,8,8-Octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane

A657881-100G A657881-25G A657881-500G A657881-5G
76727-402EA 49.87 USD
76727-402 76727-404 76727-406 76727-408
Octaphenylcyclotetrasiloxane 98%
Octaphenylcyclotetrasiloxane

Precaución: for Research Use Only

Formula: C₄₈H₄₀O₄Si₄
MW: 793.19 g/mol
MDL Number: MFCD00003268
CAS Number: 546-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR