4-Cyanophenyl-trans-4-n-propylcyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A657813-1G A657813-25G A657813-5G
76727-400EA 5 USD
76727-400 77880-381 77880-382
4-Cyanophenyl-trans-4-n-propylcyclohexanecarboxylate 97%
4-Cyanophenyl-trans-4-n-propylcyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO₂
MDL Number: MFCD09750936
CAS Number: 62439-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR