Methyl linoleate 99%

Supplier: Ambeed
Warning

Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester

A657540-25G A657540-5G A657540-100G
76727-390EA 47 USD
76727-390 76727-392 77573-118
Methyl linoleate 99%
Methyl linoleate

Precaución: for Research Use Only

Formula: C₁₉H₃₄O₂
MDL Number: MFCD00009534
CAS Number: 112-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR