S-Propyl thioacetate 98+%

Supplier: Ambeed
Danger

Synonyms: S-Propyl ethanethioate

A656146-25G A656146-5G
76727-372EA 15 USD
76727-372 76727-374
S-Propyl thioacetate 98+%
S-Propyl thioacetate

Precaución: for Research Use Only

Formula: C₅H₁₀OS
MDL Number: MFCD00039937
CAS Number: 2307-10-0
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR