Molindone hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Ethyl-2-methyl-5-(morpholinomethyl)-6,7-dihydro-1H-indol-4(5H)-one hydrochloride, 3-Ethyl-6,7-dihydro-2-methyl-5-(morpholinomethyl)-1H-indol-4(5H)-one Hydrochloride, 3-Ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-4H-indol-4-one Hydrochloride

A653237-10MG A653237-1G A653237-25G A653237-25MG A653237-50MG A653237-5G
76727-356EA 5 USD
76727-356 76727-358 76727-360 76727-362 76727-364 76727-366
Molindone hydrochloride 98%
Molindone hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂
MW: 312.84 g/mol
MDL Number: MFCD01718304
CAS Number: 15622-65-8
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR