Ni-(S)-BPB-Gly 98%

Supplier: Ambeed
Warning

A652979-100MG A652979-1G
76727-352EA 9.55 USD
76727-352 76727-354
Ni-(S)-BPB-Gly 98%
Ni-(S)-BPB-Gly

Precaución: for Research Use Only

Formula: C₂₇H₂₅N₃NiO₃
MW: 498.19 g/mol
MDL Number: MFCD12545921
CAS Number: 96293-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR