3'-Fluorobiphenyl-4-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3'-Fluoro-[1,1'-biphenyl]-4-carbaldehyde

A650345-100MG A650345-1G A650345-250MG A650345-5G
76727-328EA 25.47 USD
76727-328 76727-330 76727-332 76727-334
3'-Fluorobiphenyl-4-carbaldehyde 95%
3'-Fluorobiphenyl-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉FO
MW: 200.21 g/mol
MDL Number: MFCD04039225
CAS Number: 400750-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR