Nitrilotris(methylenephosphonic acid) 50% in water

Supplier: Ambeed
Warning

Synonyms: Tris(phosphonomethyl)amine, Nitrilotri(methylphosphonic acid)

A646401-1000G A646401-100G A646401-25G A646401-500G
76727-308EA 30.77 USD
76727-308 76727-310 76727-312 76727-314
Nitrilotris(methylenephosphonic acid) 50% in water
Nitrilotris(methylenephosphonic acid)

Precaución: for Research Use Only

Formula: C₃H₁₂NO₉P₃
MW: 299.05 g/mol
MDL Number: MFCD00002138
CAS Number: 6419-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR