Methyl-p-tolylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(p-tolyl)acetate, 4-Methylphenylacetic Acid Methyl Ester, Methyl 4-Methylphenylacetate, p-Tolylacetic Acid Methyl Ester

A646219-1G A646219-25G A646219-5G
76727-302EA 6.37 USD
76727-302 76727-304 76727-306
Methyl-p-tolylacetate 98%
Methyl-p-tolylacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
MDL Number: MFCD00056243
CAS Number: 23786-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR