Safranine O BS

Supplier: Ambeed
Warning

Synonyms: CI 50240

A645497-100G A645497-25G A645497-500G A645497-1000G
76727-290EA 15.92 USD
76727-290 76727-292 76727-294 77669-770
Safranine O BS
Safranine O

Precaución: for Research Use Only

Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR