(R)-1-(Boc-amino)-2-propanol 97% (by GC)

Supplier: Ambeed
Warning

A645295-100G A645295-1G A645295-25G A645295-5G A645295-500G
76727-282EA 131 USD
76727-282 76727-284 76727-286 76727-288 77880-365
(R)-1-(Boc-amino)-2-propanol 97% (by GC)
(R)-1-(Boc-amino)-2-propanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
MDL Number: MFCD04974338
CAS Number: 119768-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR