2'-Methoxyacetoacetanilide 98%

Supplier: Ambeed
Warning

A644702-1000G A644702-100G A644702-25G A644702-500G
76727-274EA 30 USD
76727-274 76727-276 76727-278 76727-280
2'-Methoxyacetoacetanilide 98%
2'-Methoxyacetoacetanilide

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00008781
CAS Number: 92-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR