Nerol 95%

Supplier: Ambeed
Warning

Synonyms: cis-3,7-Dimethyl-2,6-octadien-1-ol

A643146-100G A643146-25G A643146-500G
76727-266EA 28 USD
76727-266 76727-268 76727-270
Nerol 95%
Nerol

Precaución: for Research Use Only

Formula: C₁₀H₁₈O
MDL Number: MFCD00063204
CAS Number: 106-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR