N,N-Dimethyl-1-naphthylamine 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl(1-naphthyl)amine, N,N-Dimethylnaphthalen-1-amine

A643023-1G A643023-5G A643023-100G A643023-25G
76727-262EA 10.61 USD
76727-262 76727-264 77669-756 77669-758
N,N-Dimethyl-1-naphthylamine 98%
N,N-Dimethyl-1-naphthylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
MDL Number: MFCD00003919
CAS Number: 86-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR