N-Formylglycine 98% (by NMR)

Supplier: Ambeed
Warning

A642872-100G A642872-25G A642872-5G
76727-256EA 189 USD
76727-256 76727-258 76727-260
N-Formylglycine 98% (by NMR)
N-Formylglycine

Precaución: for Research Use Only

Formula: C₃H₅NO₃
MDL Number: MFCD00037360
CAS Number: 2491-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR