2,2'-Diamino-N-methyldiethylamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
N-(2-Aminoethyl)-N-methyl-1,2-ethanediamine, N,N-Bis(2-aminoethyl)methylamine
A640890-1G
A640890-5G
76727-246EA
22.28
USD
76727-246
77669-740
2,2'-Diamino-N-methyldiethylamine 97%
2,2'-Diamino-N-methyldiethylamine
Precaución: for Research Use Only
Formula:
C₅H₁₅N₃ MW: 117.20 g/mol |
MDL Number:
MFCD00198039 CAS Number: 4097-88-5 UN: 2735 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...