Potassium-4-(methoxycarbonyl)phenyltrifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(4-(methoxycarbonyl)phenyl)borate

A638924-1G A638924-250MG
76727-218EA 34 USD
76727-218 76727-220
Potassium-4-(methoxycarbonyl)phenyltrifluoroborate 98%
Potassium 4-(methoxycarbonyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₈H₇BF₃KO₂
MDL Number: MFCD04115741
CAS Number: 705254-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR