4'-tert-Butylacetophenone 98%

Supplier: Ambeed
Danger

A637396-1G A637396-25G A637396-5G
76727-202EA 10 USD
76727-202 76727-204 76727-206
4'-tert-Butylacetophenone 98%
4'-tert-Butylacetophenone

Precaución: for Research Use Only

Formula: C₁₂H₁₆O
MDL Number: MFCD00017256
CAS Number: 943-27-1
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR