Potassium-1-carboxyvinyl hydrogenphosphate 95%

Supplier: Ambeed
Warning

Synonyms: Phosphoenolpyruvate potassium salt, Potassium 1-carboxyvinyl hydrogenphosphate, Phosphoenolpyruvic acid monopotassium salt, Phosphoenolpyruvic acid potassium

A637267-250MG A637267-1G A637267-25G A637267-5G
76727-200EA 18.04 USD
76727-200 77602-612 77669-730 77669-732
Potassium-1-carboxyvinyl hydrogenphosphate 95%
Potassium-1-carboxyvinyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₃H₄KO₆P
MW: 206.131 g/mol
MDL Number: MFCD00044476
CAS Number: 4265-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR