Nickel acetate tetrahydrate 99%
Supplier: Ambeed
|
Danger
|
A635826-500G
A635826-1000G
76727-192EA
5.31
USD
76727-192
77669-728
77880-345
Nickel acetate tetrahydrate 99%
Nickel acetate tetrahydrate
Precaución: for Research Use Only
Formula:
C₄H₆NiO₄·4H₂O |
MDL Number:
MFCD00066973 CAS Number: 6018-89-9 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...